siRNA Details
| Gene Symbol: Igfbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008048.3 | NP_032074.3 | 1, 2 | 507 | ||
| GenBank mRNA | AB012886.1 | - | 1, 2 | 508 | ||
| AK002277.1 | - | 1, 2 | 503 | |||
| AK148749.1 | - | 1, 2 | 504 | |||
| AK156315.1 | - | 1, 2 | 509 | |||
| AW318969.1 | - | 1, 2 | 502 | |||
| BC047202.1 | - | 1, 2 | 419 | |||
| BC092538.1 | - | 1, 2 | 503 | 30284911 | ||
| L75822.2 | - | 1, 2 | 486 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001013048, BC086582.

Igfbp7
Genomic Map