siRNA Details
| Gene Symbol: Stau2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111272.1 | NP_001104742.1 | 11 | 1506 | ||
| NM_025303.3 | NP_079579.2 | 12 | 1637 | |||
| GenBank mRNA | AF459099.1 | - | 12 | 1587 | ||
| AF459100.1 | - | 11 | 1456 | |||
| AK015087.1 | - | 5 | 586 | |||
| AK031102.1 | - | 12 | 1564 | |||
| AK171076.1 | - | 12 | 1604 | |||
| BC010300.1 | - | 9 | 1312 | |||
| BC025118.1 | - | 11 | 1461 | 3670067 | ||

Stau2
Genomic Map