siRNA Details
| Gene Symbol: Stau2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111272.1 | NP_001104742.1 | 8 | 1057 | ||
| NM_025303.3 | NP_079579.2 | 9 | 1188 | |||
| GenBank mRNA | AF459099.1 | - | 9 | 1138 | ||
| AF459100.1 | - | 8 | 1007 | |||
| AK015087.1 | - | 2 | 137 | |||
| AK031102.1 | - | 9 | 1115 | |||
| AK171076.1 | - | 9 | 1154 | |||
| BC010300.1 | - | 6 | 863 | |||
| BC025118.1 | - | 8 | 1012 | 3670067 | ||

Stau2
Genomic Map