siRNA Details
| Gene Symbol: Olfr156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019474.2 | NP_062347.2 | 1 | 660 | ||
| GenBank mRNA | BC101957.1 | - | 1 | 660 | 40044987 | |
| BC101961.1 | - | 1 | 660 | 40044981 | ||
| BC103551.1 | - | 1 | 660 | 40044986 | ||
| BC104727.1 | - | 1 | 660 | 40044983 | ||
| Gene Symbol: Olfr157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019475.3 | NP_062348.1 | 1 | 615 | ||
| GenBank mRNA | BC137911.1 | - | 1 | 612 | 8860930 | |
| BC137912.1 | - | 1 | 612 | 8860931 | ||

Olfr156
Genomic Map