siRNA Details
| Gene Symbol: Rnf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013876.3 | NP_038904.1 | 2 | 515 | ||
| GenBank mRNA | AB024427.1 | - | 3 | 380 | ||
| AF220206.1 | - | 2 | 269 | |||
| AK014254.1 | - | 2 | 669 | |||
| AK183757.1 | - | 1 | 130 | |||
| AK210997.1 | - | 1 | 130 | |||
| BC010299.1 | - | 2 | 515 | 3155804 | ||
| BC028255.1 | - | 2 | 696 | 4972336 | ||

Rnf11
Genomic Map