siRNA Details
| Gene Symbol: Scmh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159630.1 | NP_001153102.1 | 13 | 1872 | ||
| NM_013883.2 | NP_038911.1 | 13 | 1872 | |||
| GenBank mRNA | AB030906.1 | - | 13 | 1771 | ||
| AB221680.1 | - | 9 | 1413 | |||
| AB221681.1 | - | 9 | 1413 | |||
| AK182691.1 | - | 1 | 130 | |||
| AK185818.1 | - | 1 | 129 | |||
| BC025478.1 | - | 4 | 2114 | |||
| BC034667.1 | - | 11 | 1582 | 5356322 | ||

Scmh1
Genomic Map