siRNA Details
| Gene Symbol: Zfp292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013889.2 | NP_038917.2 | 8 | 6458 | ||
| GenBank mRNA | AB093243.1 | - | 1 | 4030 | ||
| AB221605.1 | - | 1 | 681 | |||
| AF017806.1 | - | 1 | 4928 | |||
| AK034951.1 | - | 1 | 435 | |||
| BC008537.1 | - | 1 | 2546 | |||
| BC064316.1 | - | 1 | 963 | |||

Zfp292
Genomic Map