siRNA Details
| Gene Symbol: Zfp292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013889.2 | NP_038917.2 | 8 | 3313 | ||
| GenBank mRNA | AB093243.1 | - | 1 | 885 | ||
| AB221604.1 | - | 1 | 1766 | |||
| AF017806.1 | - | 1 | 1783 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp292| Gene Symbol: Zfp292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013889.2 | NP_038917.2 | 8 | 3313 | ||
| GenBank mRNA | AB093243.1 | - | 1 | 885 | ||
| AB221604.1 | - | 1 | 1766 | |||
| AF017806.1 | - | 1 | 1783 | |||