siRNA Details
| Gene Symbol: Reg3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013893.2 | NP_038921.2 | 4 | 377 | ||
| GenBank mRNA | AB028625.1 | - | 4 | 383 | ||
| AB035204.1 | - | 4 | 357 | |||
| AI594831.1 | - | 4 | 377 | |||
| AK019033.1 | - | 4 | 356 | |||
| BC113760.1 | - | 4 | 372 | 40055351 | ||

Reg3d
Genomic Map