siRNA Details
| Gene Symbol: Timm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024853.1 | NP_001020024.1 | 6 | 524 | ||
| NM_001024854.1 | NP_001020025.1 | 4 | 340 | |||
| NM_001286203.2 | NP_001273132.1 | 5 | 426 | |||
| NM_013896.3 | NP_038924.1 | 6 | 562 | |||
| GenBank mRNA | AF150101.1 | - | 5 | 385 | ||
| AK018764.1 | - | 4 | 340 | |||
| AK165848.1 | - | 3 | 2116 | |||
| BC024370.1 | - | 6 | 535 | 4976059 | ||
| CK626183.1 | - | 4 | 322 | |||
| CO799389.1 | - | 5 | 426 | |||
| CO809703.1 | - | 493 | ||||
| DV652207.1 | - | 6 | 501 | |||

Timm9
Genomic Map