siRNA Details
| Gene Symbol: Timm8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013897.2 | NP_038925.1 | 2 | 204 | ||
| GenBank mRNA | AA204614.1 | - | 2 | 193 | ||
| AF196314.1 | - | 193 | ||||
| AK003382.1 | - | 2 | 163 | |||
| AK004190.1 | - | 193 | ||||
| AK012925.1 | - | 2 | 171 | |||
| AK018718.1 | - | 2 | 171 | |||
| AK019422.1 | - | 2 | 179 | |||
| AK169258.1 | - | 2 | 183 | |||
| BC029239.1 | - | 2 | 157 | 4237232 | ||

Timm8b
Genomic Map