siRNA Details
| Gene Symbol: Timm8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013898.2 | NP_038926.1 | 1 | 98 | ||
| GenBank mRNA | AB031055.1 | - | 1 | 98 | ||
| AF150081.1 | - | 1 | 89 | |||
| AK011402.1 | - | 1 | 133 | |||
| AK088903.1 | - | 1 | 91 | |||
| AK090079.1 | - | 1 | 137 | |||
| AK165573.1 | - | 1 | 93 | |||
| BC004796.1 | - | 1 | 86 | 3588797 | ||
| BC094631.1 | - | 1 | 107 | 3419065 | ||

Timm8a1
Genomic Map