siRNA Details
| Gene Symbol: Mmp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013903.2 | NP_038931.1 | 10 | 1409 | ||
| GenBank mRNA | AF155933.1 | - | 10 | 1409 | ||
| BC152335.1 | - | 10 | 1413 | 40142709 | ||
| BC152336.1 | - | 10 | 1413 | 40142712 | ||

Mmp20
Genomic Map