siRNA Details
| Gene Symbol: Mmp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013903.2 | NP_038931.1 | 5 | 763 | ||
| GenBank mRNA | AF155933.1 | - | 5 | 763 | ||
| BC152335.1 | - | 5 | 767 | 40142709 | ||
| BC152336.1 | - | 5 | 767 | 40142712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp20| Gene Symbol: Mmp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013903.2 | NP_038931.1 | 5 | 763 | ||
| GenBank mRNA | AF155933.1 | - | 5 | 763 | ||
| BC152335.1 | - | 5 | 767 | 40142709 | ||
| BC152336.1 | - | 5 | 767 | 40142712 | ||