siRNA Details
| Gene Symbol: Kdm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013910.2 | NP_038938.1 | 3, 4 | 564 | ||
| GenBank mRNA | AK031626.1 | - | 3, 4 | 529 | ||
| AK129479.1 | - | 14, 15 | 2001 | |||
| BC057622.1 | - | 3, 4 | 559 | 6402440 | ||
| BC065090.1 | - | 14, 15 | 2200 | 6851421 | ||
| BC068239.1 | - | 11, 12 | 1754 | |||
| BU841175.1 | - | 596 | ||||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001100679.

Kdm2b
Genomic Map