siRNA Details
| Gene Symbol: Kdm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013910.2 | NP_038938.1 | 11 | 2209 | ||
| GenBank mRNA | AF176524.2 | - | 4 | 630 | ||
| AK012952.1 | - | 1 | 76 | |||
| AK031626.1 | - | 11 | 2175 | |||
| AK034033.1 | - | 5 | 1203 | |||
| AK129479.1 | - | 22 | 3646 | |||
| BC057622.1 | - | 11 | 2204 | 6402440 | ||
| BC065090.1 | - | 22 | 3845 | 6851421 | ||

Kdm2b
Genomic Map