siRNA Details
| Gene Symbol: Snai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013914.2 | NP_038942.1 | 2 | 390 | ||
| GenBank mRNA | AF133714.1 | - | 2 | 312 | ||
| AF195655.1 | - | 2 | 312 | |||
| AK042234.1 | - | 2 | 390 | |||
| AK089285.1 | - | 2 | 379 | |||
| AK153633.1 | - | 2 | 432 | |||
| AK154585.1 | - | 1 | 68 | |||
| BC100724.1 | - | 2 | 339 | 40045286 | ||
| BC100727.1 | - | 2 | 339 | 40045283 | ||

Snai3
Genomic Map