siRNA Details
Gene Symbol: Zbtb18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012330.1 | NP_001012330.1 | 2 | 584 | ||
NM_013915.3 | NP_038943.3 | 1 | 1690 | |||
GenBank mRNA | AF140224.1 | - | 1 | 1692 | ||
AK147200.1 | - | 2 | 541 | |||
BC050909.1 | - | 2 | 611 | |||
BC054529.1 | - | 1 | 692 | 5721393 | ||
BC054742.1 | - | 1 | 692 | 5721393 | ||
BC089333.1 | - | 2 | 592 | |||
BU701724.1 | - | 1 | 692 | |||
CF539422.1 | - | 2 | 592 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
