siRNA Details
| Gene Symbol: Tor1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144884.2 | NP_659133.1 | 2 | 319 | ||
| GenBank mRNA | AJ298841.1 | - | 2 | 274 | ||
| AK038891.1 | - | 2 | 276 | |||
| AK079728.1 | - | 1 | 591 | |||
| AK082936.1 | - | 2 | 229 | |||
| AK160361.1 | - | 2 | 275 | |||
| BC017683.1 | - | 2 | 299 | 4238480 | ||
| BY724297.1 | - | 2 | 308 | |||

Tor1a
Genomic Map