siRNA Details
| Gene Symbol: Tor1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133673.3 | NP_598434.2 | 2 | 419 | ||
| GenBank mRNA | AJ297743.1 | - | 2 | 449 | ||
| AK017290.1 | - | 2 | 438 | |||
| AK028859.1 | - | 2 | 233 | |||
| AK054404.1 | - | 2 | 450 | |||
| AK149754.1 | - | 2 | 440 | |||
| AK159843.1 | - | 2 | 418 | |||
| AK169692.1 | - | 2 | 442 | |||
| BC018456.1 | - | 2 | 418 | 3582246 | ||

Tor1b
Genomic Map