siRNA Details
| Gene Symbol: Pttg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131054.1 | NP_001124526.1 | 5 | 495 | ||
| NM_013917.2 | NP_038945.2 | 4 | 747 | |||
| GenBank mRNA | AF071209.1 | - | 4 | 747 | ||
| AK002473.1 | - | 4 | 461 | |||
| AK008704.1 | - | 4 | 462 | |||
| AK088387.1 | - | 4 | 461 | |||
| BC023324.1 | - | 5 | 501 | 4946234 | ||
| CN837307.1 | - | 5 | 500 | |||
| DQ157177.1 | - | 4 | 436 | |||
| DV657646.1 | - | 4 | 465 | |||

Pttg1
Genomic Map