siRNA Details
| Gene Symbol: KEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000420.2 | NP_000411.1 | 8 | 1012 | ||
| XM_005249993.2 | XP_005250050.1 | 8 | 863 | |||
| XM_005249994.3 | XP_005250051.1 | 2 | 163 | |||
| GenBank mRNA | AK314831.1 | - | 8 | 913 | ||
| BC003135.1 | - | 8 | 1012 | 2906155 | ||
| BC050639.1 | - | 8 | 907 | 5742350 | ||
| FN998902.1 | - | 7 | 802 | |||
| HE572750.1 | - | 7 | 802 | |||
| M64934.2 | - | 8 | 922 | |||

KEL
Genomic Map