siRNA Details
| Gene Symbol: Usp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013919.4 | NP_038947.2 | 2 | 406 | ||
| GenBank mRNA | AF177759.1 | - | 1 | 205 | ||
| AK011148.1 | - | 2 | 407 | |||
| AK013252.1 | - | 2 | 401 | |||
| AK049649.1 | - | 1 | 187 | |||
| BC021903.1 | - | 2 | 396 | 3985792 | ||
| KF646669.1 | - | 1 | 185 | |||

Usp21
Genomic Map