siRNA Details
| Gene Symbol: Hnf4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013920.2 | NP_038948.1 | 10 | 1498 | ||
| GenBank mRNA | AJ242626.1 | - | 10 | 1450 | ||
| AK143816.1 | - | 10 | 1499 | |||
| BC125519.1 | - | 10 | 1320 | 40130034 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnf4g| Gene Symbol: Hnf4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013920.2 | NP_038948.1 | 10 | 1498 | ||
| GenBank mRNA | AJ242626.1 | - | 10 | 1450 | ||
| AK143816.1 | - | 10 | 1499 | |||
| BC125519.1 | - | 10 | 1320 | 40130034 | ||