siRNA Details
| Gene Symbol: Prss30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013921.3 | NP_038949.2 | 3, 4 | 299 | ||
| GenBank mRNA | AJ243866.1 | - | 3, 4 | 262 | ||
| BC010970.1 | - | 3, 4 | 292 | 4216644 | ||
| BC040348.1 | - | 3, 4 | 299 | 4206523 | ||

Prss30
Genomic Map