siRNA Details
| Gene Symbol: Rnf19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013923.2 | NP_038951.1 | 5, 6 | 1513 | ||
| GenBank mRNA | AF120206.1 | - | 1406 | |||
| AF120207.1 | - | 4, 5 | 1406 | |||
| AK082665.1 | - | 5, 6 | 1465 | |||
| AK147768.1 | - | 5, 6 | 1501 | |||
| BC040769.1 | - | 4, 5 | 1288 | 4457807 | ||
| X71642.1 | - | 1, 2 | 60 | |||

Rnf19a
Genomic Map