siRNA Details
| Gene Symbol: Cbx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013926.1 | NP_038954.1 | 5 | 578 | ||
| GenBank mRNA | AB221673.1 | - | 5 | 515 | ||
| AF180370.1 | - | 5 | 578 | |||
| BC014815.1 | - | 5 | 528 | 4456898 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbx8| Gene Symbol: Cbx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013926.1 | NP_038954.1 | 5 | 578 | ||
| GenBank mRNA | AB221673.1 | - | 5 | 515 | ||
| AF180370.1 | - | 5 | 578 | |||
| BC014815.1 | - | 5 | 528 | 4456898 | ||