siRNA Details
| Gene Symbol: Cngb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013927.2 | NP_038955.1 | 6 | 853 | ||
| GenBank mRNA | AJ243572.1 | - | 6 | 853 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cngb3| Gene Symbol: Cngb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013927.2 | NP_038955.1 | 6 | 853 | ||
| GenBank mRNA | AJ243572.1 | - | 6 | 853 | ||