siRNA Details
| Gene Symbol: Aass | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013930.4 | NP_038958.2 | 9 | 1054 | ||
| GenBank mRNA | AJ224761.1 | - | 8 | 945 | ||
| AK044323.1 | - | 9 | 1035 | |||
| AK098141.1 | - | 9 | 995 | |||
| AK136216.1 | - | 9 | 1003 | |||
| AK149400.1 | - | 9 | 979 | |||
| BC005420.1 | - | 9 | 1040 | 3600654 | ||

Aass
Genomic Map