siRNA Details
| Gene Symbol: KIF2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098511.2 | NP_001091981.1 | 18 | 2087 | ||
| NM_001243952.1 | NP_001230881.1 | 18 | 2176 | |||
| NM_001243953.1 | NP_001230882.1 | 17 | 1916 | |||
| NM_004520.4 | NP_004511.2 | 17 | 1973 | |||
| GenBank mRNA | AK297347.1 | - | 18 | 1833 | ||
| AK302270.1 | - | 17 | 1610 | |||
| AK315866.1 | - | 17 | 1600 | |||
| AY317140.1 | - | 15 | 1504 | |||
| AY429552.1 | - | 4 | 244 | |||
| BC031828.2 | - | 16 | 1598 | 4932771 | ||
| EF560716.1 | - | 18 | 1798 | |||
| EF560728.1 | - | 17 | 1678 | |||
| Y08319.1 | - | 16 | 1599 | |||

KIF2A
Genomic Map