siRNA Details
| Gene Symbol: Nox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285833.1 | NP_001272762.1 | 16 | 1628 | ||
| NM_001285835.1 | NP_001272764.1 | 14 | 1511 | |||
| NM_015760.5 | NP_056575.1 | 14 | 1515 | |||
| GenBank mRNA | AB041034.1 | - | 14 | 1366 | ||
| AB042745.1 | - | 14 | 1366 | |||
| AF218723.1 | - | 14 | 1366 | |||
| AF261944.1 | - | 14 | 1370 | |||
| AF276957.1 | - | 14 | 1377 | |||
| AK050371.1 | - | 14 | 1370 | |||
| AK052905.1 | - | 14 | 1366 | |||
| AK085509.1 | - | 14 | 1371 | |||
| AK149398.1 | - | 14 | 1365 | |||
| AK149455.1 | - | 14 | 1366 | |||
| AK149492.1 | - | 14 | 1366 | |||
| BC021378.1 | - | 16 | 1476 | |||
| BC138918.1 | - | 14 | 1362 | 8861940 | ||
| BC145462.1 | - | 14 | 1358 | |||
| EU861989.1 | - | 5 | 397 | |||

Nox4
Genomic Map