siRNA Details
| Gene Symbol: Hspa14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015765.2 | NP_056580.2 | 5 | 418 | ||
| GenBank mRNA | AK031489.1 | - | 357 | |||
| AK145937.1 | - | 5 | 372 | |||
| AK153397.1 | - | 5 | 380 | |||
| BC002056.1 | - | 5 | 346 | 3493119 | ||
| BU505607.1 | - | 2 | 69 | |||
| U08215.1 | - | 5 | 378 | |||

Hspa14
Genomic Map