siRNA Details
| Gene Symbol: Ercc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015769.2 | NP_056584.2 | 1, 2 | 411 | ||
| GenBank mRNA | AF189285.1 | - | 1, 2 | 411 | ||
| AK018375.1 | - | 1, 2 | 231 | |||
| AK043366.1 | - | 1, 2 | 213 | |||
| AK138588.1 | - | 1, 2 | 205 | |||
| AK149326.1 | - | 1, 2 | 244 | |||
| BC026792.1 | - | 1, 2 | 202 | 4911382 | ||

Ercc4
Genomic Map