siRNA Details
| Gene Symbol: Sall2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244916.1 | NP_001231845.1 | 2 | 337 | ||
| NM_015772.3 | NP_056587.2 | 2 | 624 | |||
| GenBank mRNA | AB093233.1 | - | 2 | 283 | ||
| AJ007396.1 | - | 2 | 715 | |||
| AK077601.1 | - | 2 | 251 | |||
| BC054527.1 | - | 2 | 319 | |||
| BC085361.1 | - | 2 | 401 | 5706710 | ||
| CB520889.1 | - | 2 | 337 | |||
| CB525158.1 | - | 2 | 406 | |||

Sall2
Genomic Map