siRNA Details
| Gene Symbol: Hist1h1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015786.3 | NP_056601.1 | 1 | 602 | ||
| GenBank mRNA | AK002321.1 | - | 602 | |||
| AK135614.1 | - | 334 | ||||
| AK155502.1 | - | 603 | ||||
| AK209931.1 | - | 1 | 201 | |||
| AK210230.1 | - | 1 | 48 | |||
| BC089604.1 | - | 602 | 6763721 | |||

Hist1h1c
Genomic Map