siRNA Details
| Gene Symbol: Hist1h1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015787.4 | NP_056602.1 | 1 | 413 | ||
| GenBank mRNA | BC089600.1 | - | 413 | 6745595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h1e| Gene Symbol: Hist1h1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015787.4 | NP_056602.1 | 1 | 413 | ||
| GenBank mRNA | BC089600.1 | - | 413 | 6745595 | ||