siRNA Details
| Gene Symbol: Fbxw14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015793.2 | NP_056608.1 | 318 | |||
| GenBank mRNA | AF176528.1 | - | 293 | |||
| AK087709.1 | - | 318 | ||||
| AK135682.1 | - | 314 | ||||
| AK135771.1 | - | 312 | ||||
| AK166611.1 | - | 315 | ||||
| AK166612.1 | - | 315 | ||||
| BC099372.1 | - | 302 | 30938910 | |||

Fbxw14
Genomic Map