siRNA Details
| Gene Symbol: Fbxw14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015793.2 | NP_056608.1 | 609 | |||
| GenBank mRNA | AK087709.1 | - | 609 | |||
| AK135682.1 | - | 605 | ||||
| AK166611.1 | - | 606 | ||||
| AK166612.1 | - | 606 | ||||
| BC099372.1 | - | 593 | 30938910 | |||
| BC138018.1 | - | 603 | 8861037 | |||
| BC145098.1 | - | 599 | 9053646 | |||

Fbxw14
Genomic Map