siRNA Details
| Gene Symbol: KIF5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004522.2 | NP_004513.1 | 20, 21 | 2711 | ||
| NR_111932.1 | 13, 14 | 1733 | ||||
| XM_011511157.2 | XP_011509459.1 | 13, 14 | 1792 | |||
| XM_017004062.1 | XP_016859551.1 | 20, 21 | 2702 | |||
| GenBank mRNA | AB011103.1 | - | 20, 21 | 2684 | ||
| AB209191.1 | - | 1, 2 | 594 | |||
| BC110287.1 | - | 13, 14 | 1663 | 5165262 | ||
| BC127772.1 | - | 17, 18 | 2035 | |||

KIF5C
Genomic Map