siRNA Details
| Gene Symbol: Tfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289507.1 | NP_001276436.1 | 3 | 591 | ||
| NM_001289509.1 | NP_001276438.1 | 5 | 610 | |||
| NM_001289511.1 | NP_001276440.1 | 4 | 468 | |||
| NM_015799.4 | NP_056614.3 | 5 | 550 | |||
| GenBank mRNA | AF207741.1 | - | 3 | 425 | ||
| AF222895.1 | - | 4 | 468 | |||
| AK004848.2 | - | 5 | 526 | |||
| AK004965.1 | - | 5 | 581 | |||
| AK155526.1 | - | 4 | 688 | |||
| AK165320.1 | - | 3 | 419 | |||
| BC013654.1 | - | 3 | 591 | 4196597 | ||

Tfr2
Genomic Map