siRNA Details
| Gene Symbol: Crim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015800.3 | NP_056615.1 | 6 | 1549 | ||
| GenBank mRNA | AF168680.1 | - | 6 | 1001 | ||
| AK053197.1 | - | 4 | 492 | |||
| BC100348.1 | - | 6 | 1334 | 30521148 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crim1| Gene Symbol: Crim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015800.3 | NP_056615.1 | 6 | 1549 | ||
| GenBank mRNA | AF168680.1 | - | 6 | 1001 | ||
| AK053197.1 | - | 4 | 492 | |||
| BC100348.1 | - | 6 | 1334 | 30521148 | ||