siRNA Details
| Gene Symbol: Crim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015800.3 | NP_056615.1 | 2 | 961 | ||
| GenBank mRNA | AF168680.1 | - | 2 | 413 | ||
| BC100348.1 | - | 2 | 746 | 30521148 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crim1| Gene Symbol: Crim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015800.3 | NP_056615.1 | 2 | 961 | ||
| GenBank mRNA | AF168680.1 | - | 2 | 413 | ||
| BC100348.1 | - | 2 | 746 | 30521148 | ||