siRNA Details
| Gene Symbol: Ppap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005452.1 | - | 5, 6 | 789 | ||
| AK049581.1 | - | 5, 6 | 832 | |||
| AK185918.1 | - | 1, 2 | 171 | |||
| BC010332.1 | - | 5, 6 | 810 | 2654393 | ||
| BC016635.1 | - | 5, 6 | 760 | |||
| BC085104.1 | - | 5, 6 | 748 | |||

Ppap2c
Genomic Map