siRNA Details
| Gene Symbol: Capn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015830.1 | NP_056645.1 | 1 | 749 | ||
| GenBank mRNA | AK079422.1 | - | 2 | 982 | ||
| AK143711.1 | - | 3 | 1388 | |||
| AK170543.1 | - | 2 | 983 | |||
| AK212570.1 | - | 1 | 190 | |||
| BC058094.1 | - | 2 | 1007 | 6824892 | ||

Capn15
Genomic Map