siRNA Details
| Gene Symbol: Park2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016694.3 | NP_057903.1 | 11, 12 | 1389 | ||
| GenBank mRNA | AB019558.1 | - | 11, 12 | 1360 | ||
| AF250293.1 | - | 11, 12 | 1389 | |||
| AK015990.1 | - | 2, 3 | 373 | |||
| BC113204.1 | - | 11, 12 | 1372 | 40047891 | ||
| FJ555554.1 | - | 9, 10 | 1142 | |||
| FJ555555.1 | - | 6, 7 | 800 | |||
| FJ555556.1 | - | 10, 11 | 1122 | |||
| FJ555557.1 | - | 10, 11 | 1247 | |||
| GU272056.1 | - | 5, 6 | 663 | |||

Park2
Genomic Map