siRNA Details
| Gene Symbol: Hmgn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016710.2 | NP_057919.2 | 6 | 585 | ||
| GenBank mRNA | AB018374.2 | - | 6 | 540 | ||
| AF213454.1 | - | 6 | 565 | |||
| AK013748.1 | - | 5 | 454 | |||
| AK131996.1 | - | 6 | 555 | |||
| AK146755.1 | - | 6 | 561 | |||
| BC021626.1 | - | 6 | 537 | 4219308 | ||
| BC083087.1 | - | 6 | 547 | 6518798 | ||

Hmgn5
Genomic Map