siRNA Details
| Gene Symbol: Irx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018885.2 | NP_061373.1 | 6 | 1755 | ||
| GenBank mRNA | AF124732.1 | - | 6 | 1460 | ||
| AK132571.1 | - | 6 | 1755 | |||
| BC126942.1 | - | 6 | 1393 | 40089986 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irx4| Gene Symbol: Irx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018885.2 | NP_061373.1 | 6 | 1755 | ||
| GenBank mRNA | AF124732.1 | - | 6 | 1460 | ||
| AK132571.1 | - | 6 | 1755 | |||
| BC126942.1 | - | 6 | 1393 | 40089986 | ||