siRNA Details
| Gene Symbol: Galns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016722.4 | NP_057931.3 | 10, 11 | 1291 | ||
| GenBank mRNA | AA793622.1 | - | 2, 3 | 211 | ||
| AF111346.1 | - | 10, 11 | 1212 | |||
| AK159592.1 | - | 10, 11 | 1268 | |||
| AK220245.1 | - | 10, 11 | 1193 | |||
| BC004002.1 | - | 10, 11 | 1145 | 3492664 | ||

Galns
Genomic Map