siRNA Details
| Gene Symbol: Galns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193645.1 | NP_001180574.1 | 2, 3 | 377 | ||
| NM_016722.4 | NP_057931.3 | 2, 3 | 377 | |||
| GenBank mRNA | AF111346.1 | - | 2, 3 | 298 | ||
| AK033946.1 | - | 2, 3 | 295 | |||
| AK159592.1 | - | 2, 3 | 354 | |||
| AK220245.1 | - | 2, 3 | 279 | |||
| BC004002.1 | - | 2, 3 | 231 | 3492664 | ||
| BQ550288.1 | - | 2, 3 | 143 | |||

Galns
Genomic Map