siRNA Details
| Gene Symbol: Hnrnpdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016690.4 | NP_057899.2 | 5 | 983 | ||
| GenBank mRNA | AB017020.1 | - | 5 | 689 | ||
| AK011767.1 | - | 5 | 672 | |||
| AK172629.1 | - | 5 | 642 | |||
| BC021374.1 | - | 5 | 641 | 5039077 | ||
| CX219512.1 | - | 4 | 358 | |||

Hnrnpdl
Genomic Map